missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3-methoxy-beta-nitrostyrene, 99%, Thermo Scientific™

Product Code. 11461667
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461667 25 g 25g
11441667 1 g 1g
11451667 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11461667 Supplier Thermo Scientific Alfa Aesar Supplier No. B25645.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63909-38-6
Molecular Formula C16H15NO4
Molecular Weight (g/mol) 285.299
MDL Number MFCD00031043
InChI Key YDGNRJKNDGBMCL-MDZDMXLPSA-N
Synonym e-1-benzyloxy-2-methoxy-4-2-nitrovinyl benzene, 4-benzyloxy-3-methoxy-omega-nitrostyrene, 4-benzyloxy-3-methoxy-beta-nitrostyrene, 1-benzyloxy-2-methoxy-4-e-2-nitroethenyl benzene, 4-benzyloxy-3-methoxy-, a-nitrostyrene, benzene, 2-methoxy-4-2-nitroethenyl-1-phenylmethoxy, 1-benzyloxy-2-methoxy-4-2-nitroethenyl benzene, 4-benzyloxy-3-methoxy-.beta.-nitrostyrene, 4-benzyloxy-3-methoxy-^b-nitrostyrene, trans-4-benzyloxy-3-methoxy-beta-nitrostyrene
PubChem CID 687207
IUPAC Name 2-methoxy-4-[(E)-2-nitroethenyl]-1-phenylmethoxybenzene
SMILES COC1=C(C=CC(=C1)C=C[N+](=O)[O-])OCC2=CC=CC=C2

Specifications

Melting Point 122°C to 124°C
Quantity 25 g
Formula Weight 285.31
Percent Purity 99%
Chemical Name or Material 4-Benzyloxy-3-methoxy-beta-nitrostyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.