accessibility menu, dialog, popup

UserName

CAS RN 63909-38-6

H3CONOOO(E/Z)

CAS RN 63909-38-6

IUPAC Name: 1-(benzyloxy)-2-methoxy-4-(2-nitroethenyl)benzene
Synonyms: 1-(benzyloxy)-2-methoxy-4-(2-nitroethenyl)benzene
Molecular Weight (g/mol): 285.3
Molecular Formula: C16H15NO4
InChi Key: YDGNRJKNDGBMCL-UHFFFAOYSA-N
SMILES: COC1=C(OCC2=CC=CC=C2)C=CC(C=C[N+]([O-])=O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results