missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopyridine, 99+%, Thermo Scientific™

A potassium channel blocker

£48.80 - £133.00

Chemical Identifiers

CAS 504-24-5
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00006439
InChI Key NUKYPUAOHBNCPY-UHFFFAOYSA-N
Synonym 4-aminopyridine, fampridine, 4-pyridinamine, dalfampridine, 4-pyridylamine, avitrol, p-aminopyridine, 4-ap, ampyra, gamma-aminopyridine
PubChem CID 1727
ChEBI CHEBI:34385
IUPAC Name pyridin-4-amine
SMILES C1=CN=CC=C1N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15434069
View Documents
Thermo Scientific Alfa Aesar
J61470.14
25g
£48.80
25g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15444069
View Documents
Thermo Scientific Alfa Aesar
J61470.22
100g
£133.00
100g
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

504-24-5
94.117
NUKYPUAOHBNCPY-UHFFFAOYSA-N
1727
pyridin-4-amine
C5H6N2
MFCD00006439
4-aminopyridine, fampridine, 4-pyridinamine, dalfampridine, 4-pyridylamine, avitrol, p-aminopyridine, 4-ap, ampyra, gamma-aminopyridine
CHEBI:34385
C1=CN=CC=C1N

Specifications

504-24-5
273°C to 274°C
Unpleasant
MFCD00006439
105782
4-aminopyridine, fampridine, 4-pyridinamine, dalfampridine, 4-pyridylamine, avitrol, p-aminopyridine, 4-ap, ampyra, gamma-aminopyridine
C1=CN=CC=C1N
94.117
CHEBI:34385
≥99%
1.26
164°C (327°F)
C5H6N2
UN2671
14,3933
NUKYPUAOHBNCPY-UHFFFAOYSA-N
pyridin-4-amine
1727
94.12
4-Aminopyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H311-H315-H319-H335
Fatal if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c

H300-H311-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: AMINOPYRIDINES

missing translation for 'einecsNumber' : 207-987-9

missing translation for 'rtecsNumber' : US1750000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.