missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopyridine, 99+%, Thermo Scientific™

A potassium channel blocker

Brand:  Thermo Scientific Alfa Aesar J61470.14

Quantity 25g

 View more versions of this product

Product Code. 15434069

  • £48.80 / 25g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

504-24-5
94.117
NUKYPUAOHBNCPY-UHFFFAOYSA-N
1727
pyridin-4-amine
C5H6N2
MFCD00006439
4-aminopyridine, fampridine, 4-pyridinamine, dalfampridine, 4-pyridylamine, avitrol, p-aminopyridine, 4-ap, ampyra, gamma-aminopyridine
CHEBI:34385
C1=CN=CC=C1N

Specifications

504-24-5
273°C to 274°C
Unpleasant
MFCD00006439
UN2671
14,3933
NUKYPUAOHBNCPY-UHFFFAOYSA-N
pyridin-4-amine
1727
94.12
4-Aminopyridine
1.26
164°C (327°F)
C5H6N2
25g
105782
4-aminopyridine, fampridine, 4-pyridinamine, dalfampridine, 4-pyridylamine, avitrol, p-aminopyridine, 4-ap, ampyra, gamma-aminopyridine
C1=CN=CC=C1N
94.117
CHEBI:34385
≥99%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.