missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-8-(trifluoromethyl)quinoline, 97%, Thermo Scientific™

Product Code. 11499300
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499300 250 mg 250mg
11409310 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499300 Supplier Thermo Scientific Alfa Aesar Supplier No. H50193.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Amino-8-(trifluoromethyl)quinoline is used to synthesis of rubber chemical, antiseptic, antipyretic, and antiperiodic properties and are used as antimalarials and for preparing other antimalarial drugs

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 243977-15-3
Molecular Formula C10H7F3N2
Molecular Weight (g/mol) 212.18
MDL Number MFCD00269346
InChI Key JTMLOHYYNHRKTF-UHFFFAOYSA-N
Synonym 8-trifluoromethyl quinolin-4-amine, 4-amino-8-trifluoromethyl quinoline, 4-amino-8-trifluoromethylquinoline, 8-trifluoromethyl-4-quinolinamine, 8-trifluoromethyl-4-quinolylamine, 8-trifluoromethyl-quinolin-4-ylamine, 8-trifluoromethyl quinolin-4-ylamine, acmc-20ai5g, 4-quinolinamine,8-trifluoromethyl
PubChem CID 2735961
IUPAC Name 8-(trifluoromethyl)quinolin-4-amine
SMILES NC1=CC=NC2=C(C=CC=C12)C(F)(F)F

Specifications

Quantity 250 mg
Formula Weight 212.18
Percent Purity 97%
Chemical Name or Material 4-Amino-8-(trifluoromethyl)quinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.