accessibility menu, dialog, popup

UserName

NFFFNH2

CAS RN 243977-15-3

IUPAC Name: 8-(trifluoromethyl)quinolin-4-amine
Synonyms: 8-(trifluoromethyl)quinolin-4-amine
Molecular Weight (g/mol): 212.18
Molecular Formula: C10H7F3N2
InChi Key: JTMLOHYYNHRKTF-UHFFFAOYSA-N
SMILES: NC1=CC=NC2=C(C=CC=C12)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results