missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-chloro-2,6-dimethylpyrimidine, 99%, Thermo Scientific™

Product Code. 11460687
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11460687 5 g 5g
11470687 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11460687

Brand: Thermo Scientific Alfa Aesar B22499.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2858-20-0
Molecular Formula C6H8ClN3
Molecular Weight (g/mol) 157.60
MDL Number MFCD00191727
InChI Key BKFCZKYCVQQMCX-UHFFFAOYSA-N
Synonym 4-amino-5-chloro-2,6-dimethylpyrimidine, timtec-bb sbb004000, 4-amino-2-chloro-6,7-dimethylpyrimidine, 5-chloro-2,6-dimethylpyrimidine-4-ylamine, acmc-209h3e, 5-chloro-2,6-dimethyl-4-pyrimidinamine, 2,4-dimethyl-5-chloro-6-aminopyrimidine, 2,6-dimethyl-4-amino-5-chloropyrimidine, 4-pyrimidinamine,5-chloro-2,6-dimethyl, 5-chloro-2,6-dimethyl-4-pyrimidinylamine
PubChem CID 593843
IUPAC Name 5-chloro-2,6-dimethylpyrimidin-4-amine
SMILES CC1=NC(C)=C(Cl)C(N)=N1

Specifications

Melting Point 164°C to 166°C
Quantity 5 g
Formula Weight 157.6
Percent Purity 99%
Chemical Name or Material 4-Amino-5-chloro-2,6-dimethylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.