accessibility menu, dialog, popup

UserName

CAS RN 2858-20-0

CH3NCH3ClH2NN

CAS RN 2858-20-0

IUPAC Name: 5-chloro-2,6-dimethylpyrimidin-4-amine
Synonyms: 5-chloro-2,6-dimethylpyrimidin-4-amine
Molecular Weight (g/mol): 157.6
Molecular Formula: C6H8ClN3
InChi Key: BKFCZKYCVQQMCX-UHFFFAOYSA-N
SMILES: CC1=NC(C)=C(Cl)C(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results