missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-iodopyridine, 97%

Product Code. 11414400
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414400 1 g 1g
11424400 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414400 Supplier Thermo Scientific Alfa Aesar Supplier No. H50027.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88511-27-7 | C5H5IN2 | 220.013 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88511-27-7
Molecular Formula C5H5IN2
Molecular Weight (g/mol) 220.013
MDL Number MFCD02663853
InChI Key ZGOCEDVVZKFHSY-UHFFFAOYSA-N
Synonym 4-amino-3-iodopyridine, 3-iodo-4-aminopyridine, 3-iodopyridin-4-ylamine, 3-iodo-pyridin-4-ylamine, 3-iodo-4-pyridylamine, 4-pyridinamine, 3-iodo, pubchem6632, 3-iodo-4-pyridineamine, 4-amino-3-iodo-pyridin, acmc-20a09a
PubChem CID 1516510
IUPAC Name 3-iodopyridin-4-amine
SMILES C1=CN=CC(=C1N)I

Specifications

Quantity 1 g
Sensitivity Air and light sensitive
Formula Weight 220.01
Percent Purity 97%
Chemical Name or Material 4-Amino-3-iodopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.