CAS RN 88511-27-7
IUPAC Name:
3-iodopyridin-4-amine
Synonyms:
3-iodopyridin-4-amine
Molecular Weight (g/mol):
220.01
Molecular Formula:
C5H5IN2
InChi Key:
ZGOCEDVVZKFHSY-UHFFFAOYSA-N
SMILES:
NC1=CC=NC=C1I
1
–
1
of
1
results
1
4-Amino-3-iodopyridine, 97%
CAS: 88511-27-7 Molecular Formula: C5H5IN2 Molecular Weight (g/mol): 220.013 MDL Number: MFCD02663853 InChI Key: ZGOCEDVVZKFHSY-UHFFFAOYSA-N Synonym: 4-amino-3-iodopyridine,3-iodo-4-aminopyridine,3-iodopyridin-4-ylamine,3-iodo-pyridin-4-ylamine,3-iodo-4-pyridylamine,4-pyridinamine, 3-iodo,pubchem6632,3-iodo-4-pyridineamine,4-amino-3-iodo-pyridin,acmc-20a09a PubChem CID: 1516510 IUPAC Name: 3-iodopyridin-4-amine SMILES: C1=CN=CC(=C1N)I