missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ 4-Acetamido-4'-((iodoacetyl)amino)Stilbene-2,2'-Disulfonic Acid, Disodium Salt

Product Code. 11544107
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11544107 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11544107 Supplier Molecular Probes™ Supplier No. A484

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

4-acetamido-4'-((iodoacetyl)amino)stilbene-2,2'-disulfonic acid, disodium salt

The iodoacetamide derivative of stilbene (4-acetamido-4'-((iodoacetyl) amino)stilbene-2,2'-disulfonic acid, disodium salt) is readily conjugated to thiols. The combination of high polarity and membrane impermeability makes this polysulfonated dye useful for determining whether thiol-containing proteins and polypeptide chains are exposed at the extracellular or cytoplsamic membrane surface. Stilbene protein adducts are charged and can be detected by gel or capillary electrophoresis.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Product Type 4-Acetamido-4'-((iodoacetyl)amino)Stilbene-2,2'-Disulfonic Acid
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Alkyl Halide, Iodoacetamide
Label or Dye Stilbene
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.