missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dinitrocatechol, TRC

Product Code. 30385644
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385644 100 mg -
30375819 10 mg 10mg
30370836 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385644 Supplier Toronto Research Chemicals Supplier No. TRCD482560100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77400-30-7
Molecular Formula C6H4N2O6
Molecular Weight (g/mol) 200.1
Synonym 4,5-Dinitropyrocatechol, 4,5-Dinitro-1,2-benzenediol, 1,2-Dihydroxy-4,5-dinitrobenzene, 4,5-Dihydroxy-1,2-dinitrobenzene
SMILES Oc1cc(c(cc1O)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H4N2O6/c9-5-1-3(7(11)12)4(8(13)14)2-6(5)10/h1-2,9-10H
Formula Weight 200.0069
Chemical Name or Material 4,5-Dinitrocatechol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.