CAS RN 77400-30-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4,5-Dinitrocatechol, TRC
CAS: 77400-30-7 Molecular Formula: C6H4N2O6 Molecular Weight (g/mol): 200.1 Synonym: 4,5-Dinitropyrocatechol,4,5-Dinitro-1,2-benzenediol,1,2-Dihydroxy-4,5-dinitrobenzene,4,5-Dihydroxy-1,2-dinitrobenzene SMILES: Oc1cc(c(cc1O)[N+](=O)[O-])[N+](=O)[O-]