missing translation for 'onlineSavingsMsg'
Learn More

4',5,7-Trihydroxyflavone, 97%

Product Code. 11368126
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368126 100 mg 100mg
11358126 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368126 Supplier Thermo Scientific Alfa Aesar Supplier No. L15041.MC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 520-36-5 | C15H10O5 | 270.24 g/mol

4',5,7-Trihydroxyflavone is a nonmutagenic flavonoid shown to inhibit cell proliferation, angiogenesis and protein kinase. Also induces apoptosis in breast cancer cells. It inhibit topoisomerase I-catalyzed DNA re-ligation and enhance gap junctional intercellular communication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′,5,7-Trihydroxyflavone is a nonmutagenic flavonoid shown to inhibit cell proliferation, angiogenesis and protein kinase. Also induces apoptosis in breast cancer cells. It inhibit topoisomerase I-catalyzed DNA re-ligation and enhance gap junctional intercellular communication.

Solubility
Soluble in dimethyl sulfoxide (27 mg/ml) or 1M potassium hydroxide (50mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 520-36-5
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00006831
InChI Key KZNIFHPLKGYRTM-UHFFFAOYSA-N
Synonym apigenin, 5,7-dihydroxy-2-4-hydroxyphenyl-4h-chromen-4-one, chamomile, spigenin, versulin, apigenol, 4',5,7-trihydroxyflavone, apigenine, c.i. natural yellow 1, 5,7,4'-trihydroxyflavone
PubChem CID 5280443
ChEBI CHEBI:18388
IUPAC Name 5,7-dihydroxy-2-(4-hydroxyphenyl)chromen-4-one
SMILES OC1=CC=C(C=C1)C1=CC(=O)C2=C(O)C=C(O)C=C2O1

Specifications

Melting Point ∽315°C (decomposition)
Quantity 100 mg
Beilstein 262620
Merck Index 14,730
Solubility Information Soluble in dimethyl sulfoxide (27mg/ml) or 1M potassium hydroxide (50mg/ml).
Formula Weight 270.24
Percent Purity 97%
Chemical Name or Material 4',5,7-Trihydroxyflavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.