missing translation for 'onlineSavingsMsg'
Learn More

4-(4-methylperhydro-1,4-diazepin-1-yl)benzonitrile, Thermo Scientific™

Product Code. 10704097
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10704097 5 g 5g
10327064 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10704097 Supplier Thermo Scientific Maybridge Supplier No. CC60316DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 166438-80-8
Molecular Formula C13H17N3
Molecular Weight (g/mol) 215.3
InChI Key LWRMQRYMWJWCLS-UHFFFAOYSA-N
Synonym 4-4-methyl-1,4-diazepan-1-yl benzonitrile, 4-4-methylperhydro-1,4-diazepin-1-yl benzonitrile, benzonitrile,4-hexahydro-4-methyl-1h-1,4-diazepin-1-yl, 4-4-methylhomopiperazin-1-yl benzonitrile, 4-4-methyl-1,4-diazaperhydroepinyl benzenecarbonitrile, 4-hexahydro-4-methyl-1h-1,4-diazepine-1-yl benzonitrile
PubChem CID 18001106
IUPAC Name 4-(4-methyl-1,4-diazepan-1-yl)benzonitrile
SMILES CN1CCCN(CC1)C2=CC=C(C=C2)C#N

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 215.3
Percent Purity 97%
Chemical Name or Material 4-(4-Methylperhydro-1, 4-diazepin-1-yl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.