accessibility menu, dialog, popup

UserName

H3CNNN

CAS RN 166438-80-8

IUPAC Name: 4-(4-methyl-1,4-diazepan-1-yl)benzonitrile
Synonyms: 4-(4-methyl-1,4-diazepan-1-yl)benzonitrile
Molecular Weight (g/mol): 215.3
Molecular Formula: C13H17N3
InChi Key: LWRMQRYMWJWCLS-UHFFFAOYSA-N
SMILES: CN1CCCN(CC1)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results