missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenyl)-1,2,3,6-tetrahydropyridine Hydrochloride, TRC

Product Code. 30402058
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402058 5 g
30398809 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402058 Supplier Toronto Research Chemicals Supplier No. TRCF5952355G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1978-61-6
Molecular Formula C11H12lFN . HCl
Molecular Weight (g/mol) 213.68
Synonym 4-(4-Fluorophenyl)-1,2,5,6-tetrahydropyridine Hydrochloride, 4-(p-Fluorophenyl)-1,2,3,6-tetrahydropyridine Hydrochloride
SMILES Cl.Fc1ccc(cc1)C2=CCNCC2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C11H12FN.ClH/c12-11-3-1-9(2-4-11)10-5-7-13-8-6-10;/h1-5,13H,6-8H2;1H
Formula Weight 213.0721
Chemical Name or Material 4-(4-Fluorophenyl)-1,2,3,6-tetrahydropyridine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.