CAS RN 1978-61-6
IUPAC Name:
hydrogen 4-(4-fluorophenyl)-1,2,3,6-tetrahydropyridine chloride
Synonyms:
hydrogen 4-(4-fluorophenyl)-1,2,3,6-tetrahydropyridine chloride
Molecular Weight (g/mol):
213.68
Molecular Formula:
C11H13ClFN
InChi Key:
CTTBTYWBXXWBMU-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].FC1=CC=C(C=C1)C1=CCNCC1
1
–
2
of
2
results
1
4-(4-Fluorophenyl)-1,2,3,6-tetrahydropyridine Hydrochloride, TRC
CAS: 1978-61-6 Molecular Formula: C11H12lFN . HCl Molecular Weight (g/mol): 213.68 Synonym: 4-(4-Fluorophenyl)-1,2,5,6-tetrahydropyridine Hydrochloride,4-(p-Fluorophenyl)-1,2,3,6-tetrahydropyridine Hydrochloride SMILES: Cl.Fc1ccc(cc1)C2=CCNCC2