missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethyl-2-pentanone, 99%, Thermo Scientific Chemicals
£101.00 - £1516.00
Chemical Identifiers
CAS | 590-50-1 |
---|---|
Molecular Formula | C7H14O |
Molecular Weight (g/mol) | 114.19 |
InChI Key | AZASWMGVGQEVCS-UHFFFAOYSA-N |
PubChem CID | 11546 |
IUPAC Name | 4,4-dimethylpentan-2-one |
SMILES | CC(=O)CC(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11964821
|
Thermo Scientific Acros
159710050 |
5 g |
£101.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11984821
|
Thermo Scientific Acros
159710250 |
25 g |
£413.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11904831
|
Thermo Scientific Acros
159711000 |
100 g |
£1516.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
590-50-1 | |
114.19 | |
11546 | |
CC(=O)CC(C)(C)C |
C7H14O | |
AZASWMGVGQEVCS-UHFFFAOYSA-N | |
4,4-dimethylpentan-2-one |
Specifications
590-50-1 | |
100.0 | |
0.8040g/mL | |
18°C | |
99% | |
C7H14O | |
1.4025 to 1.4045 | |
0.804 | |
AZASWMGVGQEVCS-UHFFFAOYSA-N | |
4,4-dimethylpentan-2-one | |
11546 | |
99% | |
4, 4-Dimethyl-2-pentanone |
98.5 | |
Colorless to Yellow | |
125.0°C to 130.0°C | |
Authentic | |
Glass bottle | |
(CH3)3CCH2COCH3 | |
01,702 | |
4,4-dimethyl-2-pentanone, 2-pentanone, 4,4-dimethyl, methyl neopentyl ketone, neopentyl methyl ketone, methyl neoamyl ketone, 4,4-dimethyl-pentan-2-one, 2-pentanone,4,4-dimethyl, acmc-1awgm, neo-c5h11coch3 | |
CC(=O)CC(C)(C)C | |
114.19 | |
114.19 | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-685-2
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4,4-Dimethyl-2-pentanone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only