missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-2-pentanone, 99%

Product Code. 11964821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11964821 5 g 5g
11984821 25 g 25g
11904831 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11964821 Supplier Thermo Scientific Acros Supplier No. 159710050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 590-50-1 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-50-1
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
InChI Key AZASWMGVGQEVCS-UHFFFAOYSA-N
PubChem CID 11546
IUPAC Name 4,4-dimethylpentan-2-one
SMILES CC(=O)CC(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8040g/mL
Boiling Point 125.0°C to 130.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)3CCH2COCH3
Refractive Index 1.4025 to 1.4045
Quantity 5 g
Beilstein 01,702
Specific Gravity 0.804
Synonym 4,4-dimethyl-2-pentanone, 2-pentanone, 4,4-dimethyl, methyl neopentyl ketone, neopentyl methyl ketone, methyl neoamyl ketone, 4,4-dimethyl-pentan-2-one, 2-pentanone,4,4-dimethyl, acmc-1awgm, neo-c5h11coch3
Formula Weight 114.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4, 4-Dimethyl-2-pentanone
Show More Show Less
compliance-icons
Product Identifier
  • 4,4-Dimethyl-2-pentanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.