missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Di-tert-butylbiphenyl, 99%

Product Code. 11486994
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486994 5 g 5g
11496994 25 g 25g
11407004 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486994 Supplier Thermo Scientific Alfa Aesar Supplier No. B21470.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1625-91-8 | C20H26 | 266.428 g/mol

It is used in the generation of 1,2-di(lithiomethyl)benzene. It is found to accept electrons from Li metal to give a radical anion which is highly effective in the conversion of alkyl halides to alkyllithiums. 4,4?-Di-tert-butylbiphenyl is used in production of homoallylic amine derivatives. It is also used in the preparation of lithium di-tert-butylbiphenylide, a radical anion, superior to sodium or lithium naphthalenides for metalation reactions. Along with lithium, 4,4?-Di-tert-butylbiphenyl catalyzes; reaction of chloromethyl ethyl ether and different carbonyl compounds to yield corresponding hydroxyethers and reductive opening of N-phenylazetidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the generation of 1,2-di(lithiomethyl)benzene. It is found to accept electrons from Li metal to give a radical anion which is highly effective in the conversion of alkyl halides to alkyllithiums. 4,4′-Di-tert-butylbiphenyl is used in production of homoallylic amine derivatives. It is also used in the preparation of lithium di-tert-butylbiphenylide, a radical anion, superior to sodium or lithium naphthalenides for metalation reactions. Along with lithium, 4,4′-Di-tert-butylbiphenyl catalyzes; reaction of chloromethyl ethyl ether and different carbonyl compounds to yield corresponding hydroxyethers and reductive opening of N-phenylazetidine.

Solubility
Solubility in toluene; almost transparency. Soluble in dioxane: 0.1 g/mL, (clear).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1625-91-8
Molecular Formula C20H26
Molecular Weight (g/mol) 266.428
MDL Number MFCD00008834
InChI Key CDKCEZNPAYWORX-UHFFFAOYSA-N
Synonym 4,4'-di-tert-butylbiphenyl, 4,4'-di-tert-butyl-1,1'-biphenyl, 4,4'-di-t-butylbiphenyl, 1-tert-butyl-4-4-tert-butylphenyl benzene, 1,1'-biphenyl, 4,4'-bis 1,1-dimethylethyl, 1-tert-butyl-4-4-tert-butyl phenyl benzene, di-t-butylbiphenyl, pubchem9048, acmc-209st2, 4,4'-di-tertbutylbiphenyl
PubChem CID 74195
IUPAC Name 1-tert-butyl-4-(4-tert-butylphenyl)benzene
SMILES CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(C)(C)C

Specifications

Melting Point 126°C to 129°C
Boiling Point 190°C to 192°C (13 mmHg)
Quantity 5 g
Beilstein 2095855
Formula Weight 266.43
Percent Purity 99%
Chemical Name or Material 4,4'-Di-tert-butylbiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.