accessibility menu, dialog, popup

UserName

CAS RN 1625-91-8

IUPAC Name: 4,4'-di-tert-butyl-1,1'-biphenyl
Synonyms: 4,4'-di-tert-butyl-1,1'-biphenyl
Molecular Weight (g/mol): 266.43
Molecular Formula: C20H26
InChi Key: CDKCEZNPAYWORX-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C1=CC=C(C=C1)C(C)(C)C
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)