missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Phenylpropyl)pyridine, 98%

Product Code. p-7021309
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
Product Code. Quantity unitSize
11460124 50 g 50g
11470124 250 g 250g
11450124 1000 g 1000g
3 options
This item is not returnable. View return policy

Product Code. 11460124

Brand: Thermo Scientific Alfa Aesar A13590.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2057-49-0 | C14H15N | 197.281 g/mol

4-(3-Phenylpropyl)pyridine is used in electrochemical processes for a novel system of microdroplets of PPP immobilized at graphite and in mesoporous ceramic electrodes, PPP has been used to investigate electrochemically driven mono-, di-, and tri-carboxylate anion transfer at the 4-(3-phenylpropyl)pyridine/aqueous electrolyte interface for a triple phase boundary system at graphite electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(3-Phenylpropyl)pyridine is used in electrochemical processes for a novel system of microdroplets of PPP immobilized at graphite and in mesoporous ceramic electrodes, PPP has been used to investigate electrochemically driven mono-, di-, and tri-carboxylate anion transfer at the 4-(3-phenylpropyl)pyridine/aqueous electrolyte interface for a triple phase boundary system at graphite electrodes.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2057-49-0
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.281
MDL Number MFCD00047458
InChI Key AQIIVEISJBBUCR-UHFFFAOYSA-N
Synonym 4-3-phenylpropyl pyridine, pyridine, 4-3-phenylpropyl, 1-phenyl-3-4-pyridyl propane, unii-yw9q68ek6b, 4-phenylpropylpyridine, yw9q68ek6b, 1-4-pyridyl-3-phenylpropane, acmc-1ccts, dsstox_cid_24869, dsstox_rid_80544
PubChem CID 74937
IUPAC Name 4-(3-phenylpropyl)pyridine
SMILES C1=CC=C(C=C1)CCCC2=CC=NC=C2

Specifications

Density 1.029
Boiling Point 320°C to 322°C
Flash Point 140°C (284°F)
Refractive Index 1.563
Quantity 50 g
Beilstein 142242
Solubility Information Difficult to mix.
Formula Weight 197.28
Percent Purity 98%
Chemical Name or Material 4-(3-Phenylpropyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.