CAS RN 2057-49-0
CAS RN 2057-49-0
IUPAC Name:
4-(3-phenylpropyl)pyridine
Synonyms:
4-phenylpropylpyridine
Molecular Weight (g/mol):
197.28
Molecular Formula:
C14H15N
InChi Key:
AQIIVEISJBBUCR-UHFFFAOYSA-N
SMILES:
C(CC1=CC=CC=C1)CC1=CC=NC=C1
1
–
1
of
1
results
1
4-(3-Phenylpropyl)pyridine, 98%
CAS: 2057-49-0 Molecular Formula: C14H15N Molecular Weight (g/mol): 197.281 MDL Number: MFCD00047458 InChI Key: AQIIVEISJBBUCR-UHFFFAOYSA-N Synonym: 4-3-phenylpropyl pyridine,pyridine, 4-3-phenylpropyl,1-phenyl-3-4-pyridyl propane,unii-yw9q68ek6b,4-phenylpropylpyridine,yw9q68ek6b,1-4-pyridyl-3-phenylpropane,acmc-1ccts,dsstox_cid_24869,dsstox_rid_80544 PubChem CID: 74937 IUPAC Name: 4-(3-phenylpropyl)pyridine SMILES: C1=CC=C(C=C1)CCCC2=CC=NC=C2