Learn More
4-(3-Phenylpropyl)pyridine, 98%
Description
4-(3-Phenylpropyl)pyridine is used in electrochemical processes for a novel system of microdroplets of PPP immobilized at graphite and in mesoporous ceramic electrodes, PPP has been used to investigate electrochemically driven mono-, di-, and tri-carboxylate anion transfer at the 4-(3-phenylpropyl)pyridine/aqueous electrolyte interface for a triple phase boundary system at graphite electrodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-(3-Phenylpropyl)pyridine is used in electrochemical processes for a novel system of microdroplets of PPP immobilized at graphite and in mesoporous ceramic electrodes, PPP has been used to investigate electrochemically driven mono-, di-, and tri-carboxylate anion transfer at the 4-(3-phenylpropyl)pyridine/aqueous electrolyte interface for a triple phase boundary system at graphite electrodes.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Density | 1.029 |
| Boiling Point | 320°C to 322°C |
| Flash Point | 140°C (284°F) |
| Refractive Index | 1.563 |
| Quantity | 50 g |
| Beilstein | 142242 |
| Solubility Information | Difficult to mix. |
| Formula Weight | 197.28 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-(3-Phenylpropyl)pyridine |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.