missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Dimethylamino)propoxy]benzaldehyde, 97%, Thermo Scientific™

Product Code. 10092414
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10092414 5 g Amber Glass Bottle 5g
10327174 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10092414 Supplier Thermo Scientific Maybridge Supplier No. CC59504DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 26934-35-0
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD00011581
InChI Key ZYOHTXKOHWSJMZ-UHFFFAOYSA-N
Synonym 4-3-dimethylamino propoxy benzaldehyde, 4-3-dimethylaminopropoxy benzaldehyde, benzaldehyde, 4-3-dimethylamino propoxy, acmc-20ae0e, 4-3-dimethylamino propoxybenzaldehyde, 4-3-dimethylaminopropoxy-benzaldehyde, 4-3-dimethylamino-propoxy-benzaldehyde, 4-3-dimethylamino-propoxy benzaldehyde
PubChem CID 546932
IUPAC Name 4-[3-(dimethylamino)propoxy]benzaldehyde
SMILES CN(C)CCCOC1=CC=C(C=C1)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 207.27
Percent Purity 97%
Chemical Name or Material 4-[3-(Dimethylamino)propoxy]benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.