accessibility menu, dialog, popup

UserName

H3CNCH3OO

CAS RN 26934-35-0

IUPAC Name: 4-[3-(dimethylamino)propoxy]benzaldehyde
Synonyms: 4-[3-(dimethylamino)propoxy]benzaldehyde
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: ZYOHTXKOHWSJMZ-UHFFFAOYSA-N
SMILES: CN(C)CCCOC1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results