missing translation for 'onlineSavingsMsg'
Learn More

4-[(3,5-Dichloro-4-methoxyphenyl)amino]-6-methoxy-7-[3-(4-methyl-1-piperazinyl)propoxy]-3-quinolinecarbonitrile, TRC

Product Code. 30369875
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369875 10 mg 10mg
30377085 50 mg 50mg
30380578 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369875 Supplier Toronto Research Chemicals Supplier No. TRCD43538010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1391063-17-4
Molecular Formula C26H29Cl2N5O3
Molecular Weight (g/mol) 530.45
Synonym Des(2,4-dichloro-5-methoxyphenyl) 3,5-Dichloro-4-methoxyphenyl Bosutinib
SMILES COc1cc2c(Nc3cc(Cl)c(OC)c(Cl)c3)c(cnc2cc1OCCCN4CCN(C)CC4)C#N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C26H29Cl2N5O3/c1-32-6-8-33(9-7-32)5-4-10-36-24-14-22-19(13-23(24)34-2)25(17(15-29)16-30-22)31-18-11-20(27)26(35-3)21(28)12-18/h11-14,16H,4-10H2,1-3H3,(H,30,31)
Chemical Name or Material 4-[(3,5-Dichloro-4-methoxyphenyl)amino]-6-methoxy-7-[3-(4-methyl-1-piperazinyl)propoxy]-3-quinolinecarbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.