CAS RN 1391063-17-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-[(3,5-Dichloro-4-methoxyphenyl)amino]-6-methoxy-7-[3-(4-methyl-1-piperazinyl)propoxy]-3-quinolinecarbonitrile, TRC
CAS: 1391063-17-4 Molecular Formula: C26H29Cl2N5O3 Molecular Weight (g/mol): 530.45 Synonym: Des(2,4-dichloro-5-methoxyphenyl) 3,5-Dichloro-4-methoxyphenyl Bosutinib SMILES: COc1cc2c(Nc3cc(Cl)c(OC)c(Cl)c3)c(cnc2cc1OCCCN4CCN(C)CC4)C#N