missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Carboxyvinyl)benzeneboronic acid, 95%, Thermo Scientific™

Product Code. p-7046656
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368336 250 mg 250mg
11348336 1 g 1g
11358336 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368336 Supplier Thermo Scientific Alfa Aesar Supplier No. L15586.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151169-68-5
Molecular Formula C9H9BO4
Molecular Weight (g/mol) 191.977
MDL Number MFCD01075704
InChI Key IEMLKNHGGSYOMP-ZZXKWVIFSA-N
Synonym e-3-4-boronophenyl acrylic acid, 4-2-carboxyvinyl phenylboronic acid, 4-boronocinnamic acid, 3-4-boronophenyl acrylic acid, 4-2-carboxyvinyl benzeneboronic acid, 4-carboxyvin-2-yl phenylboronic acid, 4-e-2-carboxyvinyl phenylboronic acid, 4-phenylpropylester boronic acid, 4-e-2-carboxyvinyl benzeneboronic acid, 2e-3-4-dihydroxyboranyl phenyl prop-2-enoic acid
PubChem CID 5288001
IUPAC Name (E)-3-(4-boronophenyl)prop-2-enoic acid
SMILES B(C1=CC=C(C=C1)C=CC(=O)O)(O)O

Specifications

Melting Point ∼190°C to 196°C
Quantity 250 mg
Beilstein 9196727
Formula Weight 191.98
Percent Purity 95%
Chemical Name or Material 4-(2-Carboxyvinyl)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.