missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(5-Ethyl-2-pyridinyl)ethoxy]benzaldehyde, TRC

Product Code. 30379981
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30379981 50 mg
30383347 500 mg
30383914 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30379981 Supplier Toronto Research Chemicals Supplier No. TRCE92558050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114393-97-4
Molecular Formula C16H17NO2
Molecular Weight (g/mol) 255.31
Synonym 4-[2-(5-Ethyl-3-pyridyl)ethoxy]benzaldehyde
SMILES CCc1ccc(CCOc2ccc(C=O)cc2)nc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H17NO2/c1-2-13-3-6-15(17-11-13)9-10-19-16-7-4-14(12-18)5-8-16/h3-8,11-12H,2,9-10H2,1H3
Formula Weight 255.1259
Chemical Name or Material 4-[2-(5-Ethyl-2-pyridinyl)ethoxy]benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.