CAS RN 114393-97-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-[2-(5-Ethyl-2-pyridinyl)ethoxy]benzaldehyde, TRC
CAS: 114393-97-4 Molecular Formula: C16H17NO2 Molecular Weight (g/mol): 255.31 Synonym: 4-[2-(5-Ethyl-3-pyridyl)ethoxy]benzaldehyde SMILES: CCc1ccc(CCOc2ccc(C=O)cc2)nc1