missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ 4-1BB/TNFRSF9/CD137 Antibody (urelumab), Novus Biologicals™

Human Monoclonal Antibody

£162.00 - £695.00

Specifications

Antigen 4-1BB/TNFRSF9/CD137
Clone urelumab
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30044861
View Documents
Novus Biologicals™
NBP3-28248-50ug
50 μg
£162.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30045242
View Documents
Novus Biologicals™
NBP3-28248-100ug
100 μg
£266.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30046114
View Documents
Novus Biologicals™
NBP3-28248-1mg
1 mg
£695.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

4-1BB/TNFRSF9/CD137
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
TNFRSF9/ 4-1BB/ CD137
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
urelumab
Monoclonal
Purified
RUO
Human
Q07011
3604
IgG4
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.