missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ 4-1BB/TNFRSF9/CD137 Antibody (urelumab), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28248-1mg
This item is not returnable.
View return policy
Description
4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| 4-1BB/TNFRSF9/CD137 | |
| Monoclonal | |
| Unconjugated | |
| Q07011 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| urelumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| 4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9 | |
| TNFRSF9/ 4-1BB/ CD137 | |
| 1 mg | |
| Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells | |
| 3604 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction