missing translation for 'onlineSavingsMsg'
Learn More

(3R)-Hydroxyquinidine, TRC

Product Code. 30392898
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392898 1 mg -
30402115 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392898 Supplier Toronto Research Chemicals Supplier No. TRCH9532401MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60761-51-5
Molecular Formula C20H24N2O3
Molecular Weight (g/mol) 340.42
Synonym (3R)-3-Hydroxy Quinidine
IUPAC Name (2R,4S,5R)-5-ethenyl-2-[(S)-hydroxy-(6-methoxyquinolin-4-yl)methyl]-1-azabicyclo[2.2.2]octan-5-ol
SMILES COc1ccc2nccc([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@]4(O)C=C)c2c1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H24N2O3/c1-3-20(24)12-22-9-7-13(20)10-18(22)19(23)15-6-8-21-17-5-4-14(25-2)11-16(15)17/h3-6,8,11,13,18-19,23-24H,1,7,9-10,12H2,2H3/t13-,18+,19-,20-/m0/s1
Formula Weight 340.1787
Chemical Name or Material (3R)-Hydroxyquinidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.