CAS RN 60761-51-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(3R)-Hydroxyquinidine, TRC
CAS: 60761-51-5 Molecular Formula: C20H24N2O3 Molecular Weight (g/mol): 340.42 Synonym: (3R)-3-Hydroxy Quinidine IUPAC Name: (2R,4S,5R)-5-ethenyl-2-[(S)-hydroxy-(6-methoxyquinolin-4-yl)methyl]-1-azabicyclo[2.2.2]octan-5-ol SMILES: COc1ccc2nccc([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@]4(O)C=C)c2c1