missing translation for 'onlineSavingsMsg'
Learn More

(3R,4R,5R)-3-(1-Ethylpropoxy)-4-hydroxy-5-[(methylsulfonyl)oxy]-1-cyclohexene-1-carboxylic Acid Ethyl Ester, TRC

Product Code. 30381433
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381433 25 mg
30382269 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381433 Supplier Toronto Research Chemicals Supplier No. TRCE92568025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 204254-92-2
Molecular Formula C15H26O7S
Molecular Weight (g/mol) 350.43
Synonym 1-Cyclohexene-1-carboxylic acid, 3-(1-ethylpropoxy)-4-hydroxy-5-[(methylsulfonyl)oxy]-, ethyl ester, (3R,4R,5R)- (9CI, ACI), 1-Cyclohexene-1-carboxylic acid, 3-(1-ethylpropoxy)-4-hydroxy-5-[(methylsulfonyl)oxy]-, ethyl ester, [3R-(3α,4α,5β)]- (ZCI), Ethyl (3R,4R,5R)-3-(1-ethylpropoxy)-4-hydroxy-5-[(methylsulfonyl)oxy]-1-cyclohexene-1-carboxylate (ACI), (3R,4R,5R)-3-(1-Ethylpropoxy)-4-hydroxy-5-[(methylsulfonyl)oxy]-1-cyclohexene-1-carboxylic acid ethyl ester
SMILES CCOC(=O)C1=C[C@@H](OC(CC)CC)[C@@H](O)[C@@H](C1)OS(=O)(=O)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H26O7S/c1-5-11(6-2)21-12-8-10(15(17)20-7-3)9-13(14(12)16)22-23(4,18)19/h8,11-14,16H,5-7,9H2,1-4H3/t12-,13-,14-/m1/s1
Formula Weight 350.1399
Percent Purity >95%
Chemical Name or Material (3R,4R,5R)-3-(1-Ethylpropoxy)-4-hydroxy-5-[(methylsulfonyl)oxy]-1-cyclohexene-1-carboxylic Acid Ethyl Ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.