CAS RN 204254-92-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(3R,4R,5R)-3-(1-Ethylpropoxy)-4-hydroxy-5-[(methylsulfonyl)oxy]-1-cyclohexene-1-carboxylic Acid Ethyl Ester, TRC
CAS: 204254-92-2 Molecular Formula: C15H26O7S Molecular Weight (g/mol): 350.43 Synonym: 1-Cyclohexene-1-carboxylic acid, 3-(1-ethylpropoxy)-4-hydroxy-5-[(methylsulfonyl)oxy]-, ethyl ester, (3R,4R,5R)- (9CI, ACI),1-Cyclohexene-1-carboxylic acid, 3-(1-ethylpropoxy)-4-hydroxy-5-[(methylsulfonyl)oxy]-, ethyl ester, [3R-(3α,4α,5β)]- (ZCI),Ethyl (3R,4R,5R)-3-(1-ethylpropoxy)-4-hydroxy-5-[(methylsulfonyl)oxy]-1-cyclohexene-1-carboxylate (ACI),(3R,4R,5R)-3-(1-Ethylpropoxy)-4-hydroxy-5-[(methylsulfonyl)oxy]-1-cyclohexene-1-carboxylic acid ethyl ester SMILES: CCOC(=O)C1=C[C@@H](OC(CC)CC)[C@@H](O)[C@@H](C1)OS(=O)(=O)C