missing translation for 'onlineSavingsMsg'
Learn More

(3R)-2,3-Dihydro-3-(2-propyn-1-ylamino)-1H-inden-1-one, TRC

Product Code. 30374046
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374046 5 mg
30380684 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30374046 Supplier Toronto Research Chemicals Supplier No. TRCD4497655MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312077-05-6
Molecular Formula C12 H11 N O
Molecular Weight (g/mol) 185.22
Synonym 1H-Inden-1-one, 2,3-dihydro-3-(2-propyn-1-ylamino)-, (3R)-, (3R)-3-(Prop-2-yn-1-ylamino)indan-1-one, (3R)-3-(Prop-2-ynylamino)-2,3-dihydroinden-1-one, (3R)-3-[(Prop-2-yn-1-yl)amino]-2,3-dihydro-1H-inden-1-one
IUPAC Name (3R)-3-(prop-2-ynylamino)-2,3-dihydroinden-1-one
SMILES O=C1C[C@@H](NCC#C)c2ccccc12

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H11NO/c1-2-7-13-11-8-12(14)10-6-4-3-5-9(10)11/h1,3-6,11,13H,7-8H2/t11-/m1/s1
Formula Weight 185.0841
Percent Purity >95%
Chemical Name or Material (3R)-2,3-Dihydro-3-(2-propyn-1-ylamino)-1H-inden-1-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.