CAS RN 1312077-05-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(3R)-2,3-Dihydro-3-(2-propyn-1-ylamino)-1H-inden-1-one, TRC
CAS: 1312077-05-6 Molecular Formula: C12 H11 N O Molecular Weight (g/mol): 185.22 Synonym: 1H-Inden-1-one, 2,3-dihydro-3-(2-propyn-1-ylamino)-, (3R)-,(3R)-3-(Prop-2-yn-1-ylamino)indan-1-one,(3R)-3-(Prop-2-ynylamino)-2,3-dihydroinden-1-one,(3R)-3-[(Prop-2-yn-1-yl)amino]-2,3-dihydro-1H-inden-1-one IUPAC Name: (3R)-3-(prop-2-ynylamino)-2,3-dihydroinden-1-one SMILES: O=C1C[C@@H](NCC#C)c2ccccc12
2
(3R)-3-(Prop-2-yn-1-ylamino)indan-1-one (3-Ketorasagiline), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.