missing translation for 'onlineSavingsMsg'
Learn More

3M™ White Protective Coverall 4520

Product Code. 12952285
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Protective Coveralls that provide protection against liquid splashes and dust.

  • Designed to offer protection against hazardous dusts and limited liquid splashes and sprays
  • Made from lightweight and breathable material for reduced heat stress and premium wearer comfort all day
  • Standard collar for use with respiratory equipment or where a hood may not be desired
  • Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
TRUSTED_SUSTAINABILITY

Specifications

Size 3X-Large
Material Polypropylene
Color White
Disposable Single-use
Cuff Style Elastic, Knit
Closure Type 2-way Zipper with Storm Flap
Seam Type Bound
Coating Anti-static
Certifications/Compliance CE, EN 13034, EN-ISO 13982-1, EN 1149-1:2006
Description White SMMMS Polypropylene suit and green back panel, Nylon on Polyester braid zipper, Neoprene Rubber elastic, Polyester cuffs and thread
Model 4520-3 XL
Size (English) Chest 49 to 52 in.
Size (English) Inseam 35 in.
Size (English) Waist 54 in.
Size (Metric) Chest 124.46 to 132.08 cm
Size (Metric) Inseam 88.9 cm
Size (Metric) Waist 137.16 cm
For Use With (Application) Insulation Installation, Asbestos and Lead Abatement, Coal Dust in Power Plants, Woodworking, Metal Polishing, Light-Duty Building Cleaning, General Powder Handling, General Industrial Clean-up and Maintenance, Machine or Vehicle Maintenance and Food Processing
Height (English) 6.33 to 6.5 ft.
Height (Metric) 194 to 200 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'