missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Hooded Protective Coveralls 4510 Series, Elastic Wrist and Ankles
Description
- Type 5/6 Coverall designed to offer cost effective protection against hazardous dusts and limited liquid splashes
- Protection against EN 14126 Biological agents
- Made of a soft lightweight non-woven with microporous laminate for splash protection, strength and breathability
- Certified protection against Type 5/6 hazardous dusts and limited liquid splashes
- 2 panel hood design, 2-way zip with storm flap, elasticated apertures reduce the risk of skin exposure
- Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | Small |
| Material | Microporous Polyethylene Laminate |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Hood Type | 2-panel |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?