missing translation for 'onlineSavingsMsg'
Learn More

3H-Tetrafluoropropionic acid, 97%

Product Code. 11444197
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444197 10 g 10g
11434197 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444197 Supplier Thermo Scientific Alfa Aesar Supplier No. A19648.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 756-09-2 | C3H2F4O2 | 146.041 g/mol

3H-Tetrafluoropropionic acid is used to produce 2,2,3,3-tetrafluoro-propionyl chloride. This reaction will need reagent benzotrichloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3H-Tetrafluoropropionic acid is used to produce 2,2,3,3-tetrafluoro-propionyl chloride. This reaction will need reagent benzotrichloride.

Solubility
Sparingly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 756-09-2
Molecular Formula C3H2F4O2
Molecular Weight (g/mol) 146.041
MDL Number MFCD00054686
InChI Key PXRROZVNOOEPPZ-UHFFFAOYSA-N
Synonym flupropanate, 3h-tetrafluoropropionic acid, 2,2,3,3-tetrafluoropropionic acid, unii-376ks3j2q6, propanoic acid, 2,2,3,3-tetrafluoro, flupropanate iso, tetrafluoropropionic acid, pxrrozvnooeppz-uhfffaoysa
PubChem CID 69789
ChEBI CHEBI:82013
IUPAC Name 2,2,3,3-tetrafluoropropanoic acid
SMILES C(C(C(=O)O)(F)F)(F)F

Specifications

Density 1.563
Boiling Point 133°C to 134°C
Refractive Index 1.321
Quantity 10 g
UN Number UN3265
Solubility Information Sparingly soluble in water.
Formula Weight 146.04
Percent Purity 97%
Chemical Name or Material 3H-Tetrafluoropropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.