accessibility menu, dialog, popup

UserName

CAS RN 756-09-2

OOFFFF

CAS RN 756-09-2

IUPAC Name: 2,2,3,3-tetrafluoropropanoate
Synonyms: 2,2,3,3-tetrafluoropropanoate
Molecular Weight (g/mol): 145.03
Molecular Formula: C3HF4O2
InChi Key: PXRROZVNOOEPPZ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C(F)(F)C(F)F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results