Learn More
Watson-Marlow Pumps 314MC Pumphead
Description
- Designed for multi-channel low-flow applications.
- For simple tube change without disturbing other channels, cassettes are pre-loaded with tubing, which is available in twenty sizes and three different materials.
- Extension pumpheads snap fit in seconds, and two MCX (five channel) or three MCX3 (three channel extension pumpheads can be added), depending on the power of the drive.
Specifications
Specifications
| Flow Rate | 53 mL/min. |
| Model | 314MCX |
| Type | Multi-channel Pump Head |
| Rotor Material | Electroless Nickel Plated Hardened Steel, Glass Filled Nylon |
| Case Material | Glass Filled Polypropylene, IXEF (Polyarylamide), Stainless Steel |