Learn More
Axygen™ 30 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 384 Tip Format
For use with Agilent/Velocity11 automation systems
Brand:  Axygen™ VT-384-31UL-R-SDL
Code : A0
Additional Details : Weight : 10.90000kg
 View more versions of this product
Product Code. 12591268
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
  • Clear polypropylene
  • Certified RNase-/DNase-free and nonpyrogenic
  • Nonpyrogenic
  • Non-filtered
  • Available in nonsterile or sterile
Specifications
Specifications
30 μL
Non-sterile
DNase, RNase, ATP, and Endotoxin Free
Rack
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST
Non-Filtered
Clear
Non-pyrogenic, used for stringent specifications under strict process controls
Automation, Round
Polypropylene