missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 30 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 384 Tip Format
GREENER_CHOICEBIOPHARMA

Product Code. 10585180 Shop All Axygen Scientific Products
Click to view available options
Clean Claims:
DNase, RNase, ATP, and Endotoxin Free
DNase, Rnase Free
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 19200
Pack of 3840
This item is not returnable. View return policy

Product Code. 10585180

Brand: Axygen™ VT38431ULRS

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Agilent/Velocity11 automation systems

Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.

  • Clear polypropylene
  • Certified RNase-/DNase-free and nonpyrogenic
  • Nonpyrogenic
  • Non-filtered
  • Available in nonsterile or sterile

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 31 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Certifications/Compliance ANSI
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.