Learn More
3-(Trifluoromethyl)benzaldehyde, 97%, Thermo Scientific Chemicals
£28.50 - £331.00
Chemical Identifiers
CAS | 454-89-7 |
---|---|
Molecular Formula | C8H5F3O |
Molecular Weight (g/mol) | 174.122 |
MDL Number | MFCD00003373 |
InChI Key | NMTUHPSKJJYGML-UHFFFAOYSA-N |
Synonym | 3-trifluoromethyl benzaldehyde, m-trifluoromethylbenzaldehyde, benzaldehyde, 3-trifluoromethyl, alpha,alpha,alpha-trifluoro-m-tolualdehyde, alpha,alpha,alpha-trifluoro-3-tolualdehyde, unii-at7h4h5u3e, a,a,a-trifluoro-m-tolualdehyde, 3-trifluoromethyl-benzaldehyde, mtf-bad, 3-formylbenzotrifluoride |
PubChem CID | 67990 |
IUPAC Name | 3-(trifluoromethyl)benzaldehyde |
SMILES | C1=CC(=CC(=C1)C(F)(F)F)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11468693
|
Thermo Scientific Alfa Aesar
A10463.06 |
5 g |
£28.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478693
|
Thermo Scientific Alfa Aesar
A10463.14 |
25 g |
£100.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11488693
|
Thermo Scientific Alfa Aesar
A10463.22 |
100 g |
£331.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
454-89-7 | |
174.122 | |
NMTUHPSKJJYGML-UHFFFAOYSA-N | |
67990 | |
C1=CC(=CC(=C1)C(F)(F)F)C=O |
C8H5F3O | |
MFCD00003373 | |
3-trifluoromethyl benzaldehyde, m-trifluoromethylbenzaldehyde, benzaldehyde, 3-trifluoromethyl, alpha,alpha,alpha-trifluoro-m-tolualdehyde, alpha,alpha,alpha-trifluoro-3-tolualdehyde, unii-at7h4h5u3e, a,a,a-trifluoro-m-tolualdehyde, 3-trifluoromethyl-benzaldehyde, mtf-bad, 3-formylbenzotrifluoride | |
3-(trifluoromethyl)benzaldehyde |
Specifications
454-89-7 | |
83°C to 86°C (30 mmHg) | |
Almond-like | |
1.465 | |
2327537 | |
3-trifluoromethyl benzaldehyde, m-trifluoromethylbenzaldehyde, benzaldehyde, 3-trifluoromethyl, alpha,alpha,alpha-trifluoro-m-tolualdehyde, alpha,alpha,alpha-trifluoro-3-tolualdehyde, unii-at7h4h5u3e, a,a,a-trifluoro-m-tolualdehyde, 3-trifluoromethyl-benzaldehyde, mtf-bad, 3-formylbenzotrifluoride | |
C1=CC(=CC(=C1)C(F)(F)F)C=O | |
174.122 | |
174.12 | |
3- (Trifluoromethyl)benzaldehyde |
1.304 | |
68°C (154°F) | |
C8H5F3O | |
MFCD00003373 | |
Air Sensitive | |
NMTUHPSKJJYGML-UHFFFAOYSA-N | |
3-(trifluoromethyl)benzaldehyde | |
67990 | |
97% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 207-228-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only