missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzaldehyde, 97%

Product Code. 11488693
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488693 100 g 100g
11468693 5 g 5g
11478693 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488693 Supplier Thermo Scientific Alfa Aesar Supplier No. A10463.22
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 454-89-7 | C8H5F3O | 174.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 454-89-7
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.122
MDL Number MFCD00003373
InChI Key NMTUHPSKJJYGML-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzaldehyde, m-trifluoromethylbenzaldehyde, benzaldehyde, 3-trifluoromethyl, alpha,alpha,alpha-trifluoro-m-tolualdehyde, alpha,alpha,alpha-trifluoro-3-tolualdehyde, unii-at7h4h5u3e, a,a,a-trifluoro-m-tolualdehyde, 3-trifluoromethyl-benzaldehyde, mtf-bad, 3-formylbenzotrifluoride
PubChem CID 67990
IUPAC Name 3-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=CC(=C1)C(F)(F)F)C=O

Specifications

Density 1.304
Boiling Point 83°C to 86°C (30 mmHg)
Flash Point 68°C (154°F)
Odor Almond-like
Refractive Index 1.465
Quantity 100 g
Beilstein 2327537
Sensitivity Air Sensitive
Formula Weight 174.12
Percent Purity 97%
Chemical Name or Material 3- (Trifluoromethyl)benzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.