missing translation for 'onlineSavingsMsg'
Learn More

[3-(trifluoromethyl)-1H-pyrazol-5-yl]methanol, TRC

Product Code. 30453030
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453030 25 mg 25mg
30456005 250 mg 250mg
30452258 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453030 Supplier Toronto Research Chemicals Supplier No. TRCT89893025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169213-73-4
Molecular Formula C5H5N2OF3
Molecular Weight (g/mol) 166.1
Synonym 5-(Trifluoromethyl)-1H-pyrazole-3-methanol, (5-Trifluoromethyl-2H-pyrazol-3-yl)-methanol, [5-(Trifluoromethyl)-1H-pyrazol-3-yl]methanol
IUPAC Name [3-(trifluoromethyl)-1H-pyrazol-5-yl]methanol
SMILES OCc1cc(n[nH]1)C(F)(F)F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C5H5F3N2O/c6-5(7,8)4-1-3(2-11)9-10-4/h1,11H,2H2,(H,9,10)
Formula Weight 166.0354
Chemical Name or Material [3-(trifluoromethyl)-1H-pyrazol-5-yl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.